Learn More
Molecular Dimensions LAMINEX HT GLASS BASE 1 MICRON
SDP
Supplier:  Molecular Dimensions MD1150100HT
Encompass
Protein Crystallography. LAMINEX HT GLASS BASE 100 MICRON (Pack of 10) For Lipidic Cubic Phase, the Laminex system is built up from lower support plates of glass or UV transparent plastic with an adhesive spacer. On to this base, glass, UV transparent plates or films can be applied to create 96 chambers for crystal growth. Laminex offers considerable advantages for viewing and imaging such crystal growth experiments since the experiment is sandwiched between planar surfaces. The optical path creates no aberrations even when using lipidic cubic phases. The Laminex system is built up from lower support plates of glass or UV transparent plastic with an adhesive spacer. On to this base, glass, UV transparent plates or films can be applied to create 96 chambers for crystal growth. Laminex bases are available with either a 200 or 100 micron thick spacer to separate the base from the cover. Specially cut SBS size Laminex glass bases.
Catalog No. NC0761054