Learn More
Molecular Dimensions LAMINEX GLASS BASE 200 MICRON
SDP
Supplier:  Molecular Dimensions MD1150
Encompass
Protein Crystallography. LAMINEX GLASS BASE 200 MICRON (Pack of 10) For Lipidic Cubic Phase, the Laminex system is built up from lower support plates of glass or UV transparent plastic with an adhesive spacer. On to this base, glass, UV transparent plates or films can be applied to create 96 chambers for crystal growth.
Catalog No. NC0005126