Learn More
PIP™ Lady Eva Reader Bifocal
Click to view available options
Magnification Power:
+1.00
+1.25
+1.50
+1.75
+2.00
+2.25
+2.50
+2.75
+3.00
Description
- Designed for the worker who needs to see up close
- Offered in increments of +0.25 diopters to give users the exact reading power of their personal glasses
- Lightweight, rimless design provides all day user comfort
- Rubber co-molded temples provide a secure fit
- Molded nose bridge design to fit the vast majority of users
- 10 Base curve lens
- Meets ANSI Z87.1+
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Frame Color | Clear, Pink |
Magnification Power | +2.50 |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
UV Protection | 0.9999 |
Warning - Prop 65 - Reproductive Harm