Learn More
Vernier™ LabQuest 2
LabQuest 2 is a standalone interface used to collect sensor data with its built-in graphing and analysis application.
Supplier: Vernier™ LABQ2
Includes: LabQuest 2
Description
- Standalone and computer sensor interface with a touch screen
- Compatible with all Vernier sensors
- Large and high-resolution screen with portrait and landscape orientations
- Fast data collection
- Rechargeable, high-capacity battery
- Compatible with Windows and Macintosh computers
- Collect, analyze, and share sensor data wirelessly with iPad, Android devices, and Chromebooks
- Free software updates
- Large, high-resolution touch screen
- Built-in sensors such as GPS and accelerometers
- Wireless connectivity with Wi-Fi and Bluetooth
- Fast data collection with 100,000 samples per second
- Real-time graphing and live sensor data display
- Powerful analysis with linear and curve fits
- Built-in periodic table, stopwatch, scientific calculator, and more
- Touch and stylus navigation for efficiency and precision
- Includes:
- LabQuest 2 interface
- 1 stylus
- Stylus tether
- Computer connection (USB) cable
- AC power adapter
- Quick-start guide
Warranty and Services
5 year warranty (1 year on battery)
Specifications
Interface | |
Educational use only |