Learn More
Epredia™ Laboratory Marking Pen, Secureline Marker II
Mark cassettes and microscope slides with the Epredia™ Laboratory Marking Pen.
Supplier: Epredia™ 2000
This item is not returnable.
View return policy
Description
- Resistant to common laboratory solvents (water, 10% neutral buffered formalin, alcohol)
- Extra fine point for precise writing
- Non-sterile
- Not made with natural rubber latex
Specifications
Laboratory Marking Pen, 10/Pk. | |
Black |