Learn More
Miele, Inc. PG 8583LD 208V Undercounter Labwasher
Supplier: Miele, Inc. 10714830
Undercounter glassware washer with DI water connection and EcoDry function and oil seals. Performance/cycle, e.g., 128 laboratory flasks, ensures efficient use of resources and provides optimum cleaning performance.Variable speed heater pumpSafety through monitoringWash pressure and spray arm monitoringConvenient drying supportEcoDry Total securityLink to process documentation possibleLow consumption values and highest level of efficiencyThanks to innovative technology, highly developed procedures, and comprehensive system solutions, Miele Professional machines achieve outstanding reprocessing results with minimum consumption of water, energy, and process chemicals.PG 8583 Laboratory Glassware washer is certified with the ACT Label. The ACT label, also known as the Environmental Impact Factor (EIF) criteria, ensures that products carrying the label are safe and sustainable.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.