Learn More
Labomed™ 1/2-Inch Video Adapter
For use with any Lx series trinocular microscope when using LABOMED iVu3000, iVu 5000, or iVu 7000.
Supplier: Labo America Inc 9126021
Description
- Works with standard C-Mount threading
Specifications
Video Adapter | |
Video adapter Lx series trinocular microscope | |
0.5 in. Video adapter |