Learn More
Labconco™ Protector™ Fiberglass Filtered Glove Boxes
Features inlet and outlet HEPA filters, 99.99% efficient on particulates
$31161.00
Specifications
Product Type | Glove Box |
---|---|
Filter Type | HEPA |
Lighting Type | Fluorescent (2 x 25 W) |
Material | Fiberglass |
Amperage | 10 A |
Requires: Supporting base
Catalog Number | Mfr. No. | Certifications/Compliance | Plug Type | Voltage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Plug Type | Voltage | Price | Quantity & Availability | |||
16-200-402
|
Labconco™
5065031 |
CE | UK Plug | 208/230 V |
Each for $31,161.00
|
|
|||
16-200-403
|
Labconco™
5065035 |
CE | Schuko Plug | 208/230 V |
Each for $31,161.00
|
|
|||
16-200-404
|
Labconco™
5065040 |
CE | China/Australia Plug | 208/230 V |
Each for $31,161.00
|
|
|||
16-200-405
|
Labconco™
5065045 |
UL, ETL, CAN/CSA | North America Plug | 208/230 V |
Each for $31,161.00
|
|
|||
Description
- One-piece molded fiberglass liner
- Clear acrylic inner and outer transfer chamber doors
- Two glove ports
- 0.3μm particulate containment <20 ng/m3
- Built-in blower with speed control capable of airflow from 15 CFM to 85 CFM
- 0.05 ppm average tracer gas levels (surpasses ASHRAE 110 standards)
- Dry powder-coated, 18 and 20 gauge steel exterior panel
- Bag-In/Bag-Out filter disposal system
- Space-saving inner and outer transfer chamber doors pivoting upward equipped with quick-latches
- Two 8 in. ID, epoxy-coated aluminum, double-grooved glove ports mounted 17 in. apart with neoprene gaskets
- Two 25-watt fluorescent lights that exceed 120 footcandles
- LCD display for real time monitoring of volume dilution rates in cubic feet per minute, air changes/minute, and main chamber pressure (inches of water)
- Interior left-hand side wall-mounted pass-through plug
- Interior right-hand side wall-mounted electrical receptacle
- With pair of neoprene gloves, 0.015 in. thick, 30 in. long
- Large viewing area, 36W x 27.7 in. H with 0.375 in. thick, laminated safety glass
- Dimensions: 63.9W x 31.0D x 45.7 in. H (162.3 x 78.7 x 116.1cm); Interior working width: 35.5 in.
Work with hazardous materials
Certifications
Exceeds ISO Class 4 conditions and achieves ISO Class 3 conditions, CE marked (International models)
Compliance
ETL listing, UL 61010-1 and CAN/CSA C22.2 No. 1010.1 conformity (North America models)
Specifications
Glove Box | |
Fluorescent (2 x 25 W) | |
10 A | |
78.7 cm | |
45.7 in. | |
50/60 Hz | |
500 lb. | |
63.9 in. | |
162.3 cm |
HEPA | |
Fiberglass | |
31 in. | |
LCD | |
116.1 cm | |
2 | |
227 kg | |
35 in. | |
88.9 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.