Learn More
Labconco™ Final Filter for WaterPro™PS
SDP
For use with WaterPro™ PS Polishing Systems.
Supplier:  Labconco™ 9092900
Catalog No. 16-107-160
Add to Cart
Description
Description
Hollow Fiber Final Filter, self-venting. Optional filter attaches to the dispensing valve or gun. Removes all particulates and bacteria that are larger than 0.2 micron in diameter.
Specifications
Specifications
WaterPro™ PS polishing station Series 90005, 90006 and 90007 model
Hollow Fiber Final Filter
0.2 kg
Product Suggestions
Product Suggestions