Learn More
Labtech Inc EV311H Rotary Evaporator
Supplier: Labtech Inc LEV3110HV
The EV311H rotary evaporator features a manual glasssware lift and LED displays. The micro lifting switch ensures easy locking and elevating. Rotation speed and bath temperature of evaporating process can be easily adjusted by controller and is shown on the digital display. Sample Volume Range: 50ml to 3000ml, Rotation speed: 20-260 rpm. 5L Heating bath supports both oil and water applications with temperature range: Ambient - 180C. Includes 1L evaporation and collection flasks and vertical condenser. Dimensions: 15L14W28H in.; Weight: 51Lbs, 115V, 60Hz.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.