Learn More
Com-Pac International™ Lab Corp Insulated Infectious Substance Shipper
Supplier: Com-Pac International™ LCCT5000
Description
United Nations (UN)-certified container provides maximum insulation performance for shipping infectious substances on dry ice. Package contains a liquid-tight, 0.5-liter, polyethylene pressure vessel. Alcohol may be use to sterilize this container. Insulation provides up to 60 hours of temperature protection using 5.6 pounds of dry ice. Shipper items include compartmental bubble pouch, absorbent sheet, all domestic and international container markings, labels and shipping declarations. Instructions for use are printed on the box flap. The pressure vessels are able to withstand 95kPa, avoiding leakage even during high-altitude transportation.
Order Info
Inside dimensions, in. (cm): 7 1/2 x 4 1/4 x 8 1/2 (19.1 x 10.8 x 21.6) with a 1/2-liter vessel. Outside dimensions, in (cm): 11 x 8 x 12 1/4 (27.9 x 20.3 x 31.1).
Compliance
Shipper has been independently tested and certified to comply with applicable UN regulations, DOT regulations and ICAO/IATA Packing Instructions 602 and 904.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.