Learn More
Com-Pac International™ Lab Corp FX-11 Infectious Substance Shipper
Supplier: Com-Pac International™ LCCT3055
Description
Special stop-lock top provides optimum security; United Nations (UN)-certified shipper; Complies with ICAO/IATA Packing Instruction 602.
Combines UN certification requirements with new innovative safety features. Shipper consists of a transparent pressure vessel able to withstand up to 95kPa, cushioning materials, outer box, labels, declarations and instructions. Transparent pressure vessel allows contents to be seen. The red stop-lock closure is designed to provide optimum security during transport; yet, opens easily with a quarter turn. The Lab Corp FX-11 Infectious Substance Shipper is specifically designed to meet the additional specifications required by Federal Express.
Order Info
Dimensions, in.: 6 x 12 x 8 with 1-liter vessel. Also available with two pressure vessels, doubling shipping capacity (22-432-19).
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.