Learn More
Lab-Aids™ Rh Blood-Typing Kit
Determine Rh factor, understand Rh mechanisms and relationships to transfusion and genetics and observe tissue compatibility.
Supplier: Lab-Aids™ 1A32
Includes: Vial of anti-Rh serum, 50 mixing spatulas, 50 presaturated alcohol pads, 50 sterile disposable blood lancets, 50 Lab-Aids Slide Guides™, complete instruction manual
Description
The significance of the Rh factor becomes clear as students type their own blood to determine if they are Rh Positive or Rh Negative. In the first part of this activity, students determine their own Rh blood type. This is done by typing their blood and observing tissue incompatibility. Next, they mix a blood sample with Rh serum. Finally, the students observe the resulting film for agglutination. If agglutination occurs, the red cells possess the Rh factor and the individual is considered to be Rh+. If agglutination doesn't occur, they are Rh-. No warming of the serum or light box is necessary. This interesting kit includes background information on the discovery of the Rh factor, the physiological basis for Rh incompatibility, and the procedure used for correcting this among newborns. The exclusive Slide Guide gives each student a permanent notebook record of the experiment. This kit contains enough material for 32 student tests.Specifications
Understand Rh mechanisms and relationships to transfusion and genetics | |
Vial of anti-Rh serum, 50 mixing spatulas, 50 presaturated alcohol pads, 50 sterile disposable blood lancets, 50 Lab-Aids Slide Guides, complete instruction manual | |
Rh Blood Typing Kit | |
32 Students |