Learn More
Lab-Aids™ Blood Clotting Experiment Kit
Learn about coagulation rates and bleeding times, use microscope to observe fibrin threads in clotting and observe the effects of calcium in blood clotting.
Supplier: Lab-Aids™ 2
Includes:
- 50 Microscope slides
- 50 Capillary tubes
- 50 Sterile, disposable blood lancets
- 50 Presaturated alcohol pads
- 50 Red mixing spatulas
- Package of absorbent paper
- Drop-controlled bottle of calcium carbonate solution
- Drop-controlled bottle of saline-citrate solution
- 50 Student worksheets and guides
- Instruction manual with MSDS
Requires : Microscopes
Description
Students will examine their own blood to better understand the clotting process. Students will discover their own clotting times studying coagulation rates and bleeding times, observe the formation of fibrin threads in clotting and study the effects of calcium on blood clotting. Individualized worksheets and sterile components make this a highly motivational lab activity. Microscopes are needed.Specifications
Understanding coagulation rates and bleeding times | |
50 Microscope slides, 50 capillary tubes, 50 sterile, disposable Blood lancets, 50 presaturated Alcohol pads, 50 red mixing spatulas, package of absorbent paper, drop controlled bottle of calcium carbonate solution, drop controlled bottle of Saline citrate solution, 50 Student worksheets and guides, Instruction manual with MSDS | |
Blood Clotting Experiment Kit | |
50 Students |