Learn More
Lab-Aids™ Bacteria Study Kit
Culture bacteria, prepare bacterial smears and stain them using Gram stain and observe/identify bacteria.
Supplier: Lab-Aids™ 160
Includes:
- All required materials
- Detailed instructions
- Background information
- 30 student guides
Description
Students conduct the classic Leeuwenhoek bacteria experiment in this individualized, self-paced kit. Paralleling methods performed over 300 years ago, students observe and identify bacteria using a microscope. Microscopes required. Prepared bacteria slides are also available if desired.Order Info
Class size: 30 students.
Specifications
Bacteria Study Kit | |
Culture bacteria, prepare slides for microscopic study and apply gram stain | |
Teacher's guide with MSDS 1, 30 student worksheets and guides, 2 droppers, 30 microscope slides, Beans Pkg, Hay Pkg, Peppercorns Pkg, 60 Plastic transfer pics, 6 transparent infusion containers, 2 Gram Iodine solution Vial, 2 Crystal Violet stain drop control bottle, 2 Methylene blue stain drop control bottle, 2 Safranin stain drop control bottle | |
30 Students |