Learn More
U.S. Pharmacopeia L-Phenylalanine, 63-91-2, MFCD00064227, 200mg
Supplier: U.S. Pharmacopeia 1530503
Molecular formula C9H11NO2, Molecular weight 165.19, Melting Point 541.4 °F (283 °C) (decomposes), Boiling Point 563 °F (295 °C) (sublimes), Synonyms: L-2-amino-3-phenylpropionic acid