Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Methionine (White Crystals or Crystalline Powder), Fisher BioReagents
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
Description
This amino acid is suitable for use in tissue culture systems requiring additives.
Specifications
Melting Point | 276°C |
Ammonium | 0.02% max. |
Arsenic (As) | 1ppm max. |
pH | 5.5 to 6.5 |
Color | White |
Heavy Metals (as Pb) | 10ppm max. |
Assay Percent Range | 98.5 to 100.5 % |
Identification | Pass Test |
Loss on Drying | 0.2% max. |
Merck Index | 15, 6047 |
Show More |
Safety and Handling
Recommended Storage | RT |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction