Learn More
U.S. Pharmacopeia L-Methionine, 63-68-3, MFCD00063097, 200mg
Supplier: U.S. Pharmacopeia 1411504
Molecular formula C5-H11-N-O2-S, Molecular weight 149.21 g/mol, Melting Point 536 - 550.4 °F (280 - 288 °C), Boiling Point 366.8 °F (186 °C)