Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Histidine, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
25 g
100 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
71-00-1
C6H9N3O2
155.16
MFCD00064315
HNDVDQJCIGZPNO-UHFFFAOYNA-N
2-amino-3-(1H-imidazol-5-yl)propanoic acid
NC(CC1=CN=CN1)C(O)=O
71-00-1
C6H9N3O2
155.16
MFCD00064315
HNDVDQJCIGZPNO-UHFFFAOYNA-N
2-amino-3-(1H-imidazol-5-yl)propanoic acid
NC(CC1=CN=CN1)C(O)=O

H1021, 71-00-1, C6H9N3O2

Spectrum™ Chemical L-Histidine, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Chloride 0.05%
Ignition Residue 0.4%
Loss on Drying 0.2%
Packaging Fiber Drum
Specific Rotation +12.6° to +14.0°
Sulfate 0.03%
Percent Purity 98.5 to 101.5%
Grade USP
Quantity 25 kg