Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Cystine (White Crystals or Cryst. Powder), Fisher BioReagents

Catalog Number BP377100
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
56-89-3
C6H12N2O4S2
240.292
LEVWYRKDKASIDU-IMJSIDKUSA-N
l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
67678
CHEBI:16283
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
C(C(C(=O)O)N)SSCC(C(=O)O)N
56-89-3
C6H12N2O4S2
240.292
LEVWYRKDKASIDU-IMJSIDKUSA-N
l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
67678
CHEBI:16283
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
C(C(C(=O)O)N)SSCC(C(=O)O)N
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY
Melting Point 260°C
Ammonium 0.02% max.
Arsenic (As) 1ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.1% max.
Assay Percent Range 98.5 to 101.0 %
Identification Pass Test
Loss on Drying 0.3% max.
Merck Index 15, 2779
Other Amino Acids Chromatographically not detectable
Packaging Glass Bottle
Specific Rotation -215 to -225° (+ or -)
Formula Weight 240.29
Percent Purity 98.5 to 101.0%
State of Solution (%T) 98.0% min.
Alpha Vector Cystine
Quantity 100 g
Physical Form Powder
Chemical Name or Material L-Cystine
Show More Show Less
Recommended Storage RT
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit