Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Arginine, Tocris Bioscience™

Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
74-79-3
C6H14N4O2
174.20
MFCD00002635
ODKSFYDXXFIFQN-UHFFFAOYNA-N
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
6322
CHEBI:16467
2-amino-5-[(diaminomethylidene)amino]pentanoic acid
NC(CCCN=C(N)N)C(O)=O
This item is not returnable. View return policy
74-79-3
C6H14N4O2
174.20
MFCD00002635
ODKSFYDXXFIFQN-UHFFFAOYNA-N
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
6322
CHEBI:16467
2-amino-5-[(diaminomethylidene)amino]pentanoic acid
NC(CCCN=C(N)N)C(O)=O

Endogenous substrate for NOS

Precursor in the formation of nitric oxide.
Formula Weight 174.2
Percent Purity >98%
Quantity 100 mg
Chemical Name or Material L-Arginine

Recommended Storage : Store at Room Temperature

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit