Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Kut-Gard™ Polyester-Wrapped Stainless Steel Core Cut-Resistant Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Description
- Seamless knit construction provides comfort, fit and dexterity
- PolyKor™ engineered yarns offer cut and slash resistance
- Ambidextrous pattern permits glove to be worn on either hand
- Special non-latex cuff dramatically reduces cuff blowout, even after multiple launderings in bleach
- Can be worn on its own or as a glove liner
- Secure gripping properties
- Shrink resistant
- Made from component materials that comply with Federal Regulations for food contact 21 CFR, 170-199
- Products constructed with PolyKor™ engineered yarn are suitable for protection against mechanical hazards in the toughest environments
- PolyKor products are highly resistant to abrasion and chemicals, so you can rely on them to provide long-lasting protection
Specifications
Certifications/Compliance | FDA 21 CFR, 170-199 |
Construction | Seamless Knit |
Coating Material | Uncoated |
Color | Blue |
Cut Resistance Rating | ANSI A5 |
Material | Stainless Steel Fiber, Synthetic Fiber |
Product Type | Cut-resistant Gloves |
Size | X-Small |
For Use With (Application) | Application where sharp cutting instruments are used, such as food service, food processing and glass/metal handling |
Glove Cuff Color | Black |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction