Learn More
REPLIGEN CORPORATION KROSFLO FITTING/CLAMP1.5IN 3/8
Supplier: REPLIGEN CORPORATION ACPXLM306N
KrosFlo® Fittings-1.5 in; 3/8in 90; KrosFlo® fittings simplify and make a Tangential Flow Filtration path more robust. The sanitary flange of the KrosFlo fittings is a standard sanitary clamp. Tubing and pressure transducers can then be attached to the KrosFlo's hose barb and female luer port, eliminating the need for tube-to-tube adapters, fittings and connections. KrosFlo fittings are molded polysulfone, the same low-leachable and highly compatible polymer used in Spectrum's hollow fiber module housings. KrosFlo fittings are available in 6 sizes, 2 with a 90 degree hose barb fitting which makes connections for some applications easier.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.