Learn More
Koehler™ Instrument Microprocessor-Based Digital Penetrometer
Microprocessor-based digital penetrometer tests the consistency of lubricating greases, petroleum waxes, bitumens, pastes, creams, food products, and other solid to semi-solid materials
Supplier: Koehler™ Instrument K9559000000
Includes:
- 47.5g standard plunger
- 50g and 100g weights
- Rechargeable, built-in battery pack or AC operation
Description
User-friendly Operation
- Separate keypad controls for each parameter
- Choice of programmable or full manual operation using coarse and fine push-button controls Can also be programmed with four sample surface height presets, automatically timed penetration measurements and test intervals, and a curing or temperature stabilization period for delayed penetrator release Memory stores selected parameters for automatic use in subsequent tests until changed by user Built-in circuit senses sample surface for automatic placement when testing electronically conductive samples
- Audible signal and visual error messages
- Test results can be communicated to a printer or computer via a built-in RS-232 interface
- Built-in magnifier and illuminator arms
- Large digital display located on head of unit displays all functions, countdown of penetration and delay intervals, and test results
- Detachable machined base with large platform accommodates many sizes of sample containers or constant temperature cylinders
- Optional Software Package provides additional features; see 14-210-36 at bottom
Compliance
Conforms to all ASTM™, IP, ISO 9001, and related specifications.
Specifications
Digital Penetrometer | |
220-240V 50/60Hz |