Learn More
Koehler™ Instrument Adjustable-Levels Sample Thief
Supplier: Koehler™ Instrument K27900
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Specifications
Sample Thief | |
16 oz. | |
Yes | |
Adjustable needle valve |
Plated Brass | |
473 mL | |
ASTM D4057, D6074 or ASTM D1265 |