Learn More
DWK Life Sciences Kimble™ Disposable Soda Lime Pasteur Pipette
The Kimble™ Disposable Soda Lime Pasteur Pipette is manufactured from 90 expansion soda lime glass conforming to USP Type III requirements.
Supplier: DWK Life Sciences 63B93
Description
- Soda lime pipets are manufactured from 90 expansion soda lime glass tubing conforming to USP Type III requirements
- Ideal for the one-time use for small volume liquid transfer
- Available either unplugged or with a cotton plug to suit most applications
Specifications
Soda Lime Glass | |
9 in. | |
2 mL | |
Unplugged Pasteur Pipet |
Non-sterile | |
Clear | |
22.86 cm | |
Single-use |