Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark™ Professional Kimberly-Clark Professional™ Universal Folded Towel Dispenser

Graceful, curved, contemporary look to create the best impression for your restrooms. Ideal for Kleenex™ and Scottfold™ Multi-folds and C-fold towels. No adaptors or inserts needed. When installed properly this dispenser meets the ADA Standards for Accessible Design; 28 CFR Part 36; local rules may vary.

Supplier:  Kimberly-Clark™ Professional 09906

Catalog No. 19-413-104

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • 1 Kimberly Clark Professional Universal Folded Paper Towel Dispenser / Case; White
  • Dispenser dimensions: 13.3 in. x 5.9 in. x 18.9 in.
  • Manual dispensing of multi fold paper towels and cfold paper towels (sometimes called trifold paper towels)
  • It's simple: no adaptors, inserts or batteries needed
  • The dispenser is ADA compliant when installed properly and when using recommended paper towels

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Universal Folded Towel Dispenser
13.31 in.
18.85 in.
ADA Standard
White
34 cm
48 cm
Ideal for scotfold towels, multi-folds and C-folds
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit