Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ KleenGuard™ G10 Arctic Blue Nitrile Gloves

Catalog Number p-3776344
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

KleenGuard™ G10 gloves provide durability and strength for light-duty tasks. Ideal for industrial, manufacturing and food processing applications.

  • Powder free, nitrile
  • Textured fingertips provide enhanced grip and dexterity
  • Ambidextrous
  • Beaded cuff
  • Puncture-resistant formulation delivers barrier protection against chemical splash, dirt, grease and oil
  • Ideal for cleaning and maintenance, manufacturing, food processing, electronic assembly and painting applications
TRUSTED_SUSTAINABILITY
Certifications/Compliance FDA 21 CFR 170-199, CFIA approved
Applications Industrial
AQL 4
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size X-Small
Thickness (English) 2 mil
Thickness (Metric) 0.05 mm
Model 90095
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit