Learn More
Kimberly-Clark Professional™ KleenGuard™ V80 SG34 Safety Goggles
Catalog Number p-3203185
Click to view available options
Description:
Anti-fog Lens
Standard lens
This item is not returnable. View return policy
This item is not returnable. View return policy
When you need comfortable splash protection goggles at an economical price point, look no further than the KleenGuard V80 SG34 Safety Goggles.
  • Clear polycarbonate lenses allow maximum visible light transmission and are ideal for indoor use
  • Button vents prevent indirect venting
  • Fit over most prescription eyeglasses
  • Headband buckles are located toward the rear of the frame to limit interference with peripheral vision on the job
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Certifications/Compliance ANSI Z87.1-2010
Description Anti-fog Lens
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Frame Color Green
Size Universal
Ventilation Details Button vents
Lens Material Polycarbonate
Frame Type Soft, pliable frame for enhanced fit and comfort