Learn More
DWK Life Sciences Kimble™ KIMAX™ Glass, Bottle Reservoir
Capacity Metric: 2 L
Supplier: DWK Life Sciences 17224F2000
Description
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirementsSpecifications
Reservoir Bottle | |
2 L | |
White enamel marking spot |
0.53 gal. | |
Glass |