Learn More
Lonza Walkersville KGM Gold Keratinocyte Growth Medium SingleQuots Supplements and Growth Factors
Supplier: Lonza Walkersville 00192152
KGM Gold SingleQuot Kit are components sold separately. These are part of the KGM Gold Keratinocyte Growth Medium BulletKit (Catalog Number 192060). The full KGM Gold BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 192060.KGM Gold BulleKit is a new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.Content:1 x Natural Cap Vial with Hydrocortisone 0.50 mL1 x Natural Cap Vial with Transferrin 0.50 mL1 x Amber Vial with Epinephrine 0.25 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Orange Cap Vial with BPE 2.00 mL1 x Green Cap Vial with hEGF 0.50 mL1 x Lilac Cap Vial with Insulin 0.50 mL
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.