Learn More
METTLER TOLEDO 3P INDUSTRIAL KF Coulometer C30SX Without diaphragm
Supplier: METTLER TOLEDO 3P INDUSTRIAL 30252662
The C30SX delivers top flexibility in the Compact KF Coulometer line. Accurate results from 1 ppm to 5% water. Supports 150 methods, LabX Software for full data management and automated InMotion KF Ovens for high sample throughputs.