Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoprofen, USP, 98.5-101%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
22071-15-4
C16H14O3
254.29
DKYWVDODHFEZIM-UHFFFAOYNA-N
2-(3-benzoylphenyl)propanoic acid
CC(C(O)=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
22071-15-4
C16H14O3
254.29
DKYWVDODHFEZIM-UHFFFAOYNA-N
2-(3-benzoylphenyl)propanoic acid
CC(C(O)=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1

K1155, 22071-15-4, C16H14O3

Spectrum™ Chemical Ketoprofen, USP is a nonsteroidal anti-inflammatory drug. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Melting Range 92.0-97.0°C
UN Number UN2811
Packaging Fiber Drum
Quantity 25 kg
Specific Rotation +1 to -1°
Percent Purity 98.5 to 101%
Grade USP