Learn More
Kemtec™ Chemistry Kit: Ionic Interaction
Introduce junior high and high school students to writing ionic formulas and ionic interaction equations.
Supplier: Aqua Phoenix Scientific 4104
Alert:
Choking hazard (1) Small parts. Not for children under 3 yrs.
Includes: Depression plates, plastic toothpicks, Calcium Nitrate, Potassium Sulfate, Cadmium Nitrate, Sodium Silicate, Cobalt Nitrate, Potassium Oxalate, Barium Nitrate, Potassium Phosphate, Cupric Chloride, Potassium Chromate, Potassium Iodide, Acetic Acid, Manganese Nitrate, Potassium Hexacyanoferrate (iii) (ferricyanide), Nickel Nitrate, Potassium Hexacyanoferrate (ii) (ferrocyanide), Strontium Nitrate, Silver Nitrate, Lead Nitrate, Ferric Nitrate, Zinc Nitrate, Chromium Nitrate, Aluminum Nitrate, Potassium Bromide, Potassium Hydroxide, Potassium Chloride, Potassium Carbonate, instructor's manual with reproducible student sheet, SDS
Requires : Goggles
Description
This kit provides an introduction to ionic interactions by giving students exposure to a large number of reactions, allowing maximal observation for a limited amount of time and effort.
- Includes precipitate forming, gas forming and non-ionized product forming reactions
- Relate placement of elements in periodic table to their reactivity
- Extend to write ionic formulas for products formed and ionic interaction equations
- Develop solubility principles from empirical evidence
- Ideal for high school chemistry or physical sciences studies
- Grades 9-12
- Class size: 24 students
Specifications
Ionic Interaction Chemistry Kit | |
7 to 14 | |
To introduce junior high and high school students to writing ionic formulas and ionic interaction equations | |
Depression plates (24 well), Packs toothpicks, 0.1M Barium Nitrate, 0.05M Calcium Nitrate, 0.05M Cadmium Nitrate, 0.1M Cobaltous Nitrate, 0.1M Cupric Chloride, 0.026M Mercuric Nitrate, 01M Manganese Nitrate 0.05M Nickel Nitrate, 0.1M Strontium Nitrate, 0.05M Lead Nitrate, 0.05M Zinc Nitrate, 0.05M Aluminum Nitrate, 0.05M Ferric Nitrate, 0.05M Chromium Nitrate, 0.05M Potassium Bromide, 0.05M Potassium Chloride, 0.05M Potassium Iodide, 0.1M Potassium Hydroxide, 0.05M Potassium Carbonate, 0.05M Potassium Sulfate, 0.05M Sodium Silicate, 0.05M Potassium Oxalate, 0.05M Potassium Phosphate, 0.05M Potassium Chromate, 0.1M Acetic Acid, 0.05M Potassium Ferricyanide (dark), 0.05M Potassium Ferrocyanide (light), 0.05M Silver Nitrate, instructor manual, student data sheets, SDS, assessment manual |