Learn More
Kimberly-Clark Professional™ KC600 Kimguard™ Sterilization Wrap
Two sheets bonded together, making it possible to double wrap in single step
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
19-077-541
|
Kimberly-Clark Professional™
62672 |
54 x 72 in. | 54 x 72 in. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Two layers of wrap, thermally sealed along the sides
- Delivers the protection of double-wrapping in a single step, in about half the time it takes to wrap or open packages using sequential double wrapping
- Quick way to ensure that instrument sterility is uncompromised
- Microbial barrier protection with patented Powerguard™ technology
- Made in USA
- Pre-Vacuum Steam and Ethylene Oxide (EO) Compatible
- Flame Resistance: Meets NFPA Class 1
- Blue