Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kawasumi™ Standard Port Access Infusion Sets - Microbore 8in. Tubing
Supplier: Kawasumi™ PI03091
Specifications
Standard Port Access Infusion Set | |
19 G | |
1 in. |
Brown | |
Female luer lock connector with flange, non coring needle, needle hub and wing | |
8 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction