Learn More
Dynalon™ Kartell™ Stackable Deep Trays
HDPE, stackable and low form trays.
$75.00 - $104.00
Description
Deep Trays
- With handles for transporting and managing spills
- Rectangular tank with draining ports on all sides
- Useful for holding defrosting samples and plant cultures
Specifications
HDPE | |
Deep Tray | |
Handle | |
Rectangular |