Learn More
Dynalon™ Kartell Bucket with Graduations and Spout
For measuring and transporting liquids or solids. Dynalon™ Kartell Bucket with Graduations and Spout are chemical resistant especially at room temperature.
Supplier: Dynalon™ 2131850009
Description
- Features steel bale handle with comfortable grip
- Made of LDPE
- Graduated internally in liters
- Use up to 80˚C or well below freezing