Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappler™ Frontline 300 Chemical and Fire Protection Jacket
Click to view available options
Closure Type:
Double Hook and Loop Storm Flap, Zipper
Hook and Loop Storm Flap, Zipper
Size:
2X-Large/3X-Large
Large/X-Large
Specifications
Size | Large/X-Large |
Material | Frontline 300 |
Color | Silver |
Closure Type | Hook and Loop Storm Flap, Zipper |
Construction | Heat Sealed and Taped Seam |
Cuff Style | Cone inserts |
Seam Type | Sealed |
For Use With (Application) | Petrochemical operations, line maintenance, tank cleaning, refueling situations |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction