Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Kangaroo Leather Super Glove™ - Gauntlet Cuff
Click to view available options
Size:
Small
2X-Large
Description
- Air Spacer™ Thermal Architecture traps air for thermal protection without adding bulk to the glove
- Black Digiroo™ Digital Kangaroo Grip on the palm and fingers for durability, grip, and tactility
- Crosstech™ Moisture Barrier is stable and durable with high temperature resistance
- Bubble-Flex Construction on the knuckles and fingers features dead-air spacer ridges for natural hand flexing
- Top Grain Kangaroo Leather provides abrasion resistance and exceptional durability
- Anthropomorphically correct 3D-style fights hand fatigue and reduces injury
Certifications
NFPA 1971-2007
Specifications
Back Material | Top Grain Kangaroo Leather |
Certifications/Compliance | NFPA 1971-2007 |
Coated Area | Palm, Fingers |
Cuff Material | Cowhide Leather |
Color | Blue |
Hand | Specific |
Product Type | Structural Firefighting Gloves |
Size | Small |
Glove Cuff Color | Black |
Glove Cuff Style | Gauntlet |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction