Learn More
YSI 3P LifeScience KAL KIT TURBITITY 550 IR & WT
Supplier: YSI 3P LifeScience 600542
Calibration kit for turbidimeter Turb 550 and 550 IR: 0.02 - 10.0 - 1000 NTU, cleaning tissues and indexing rings.