Learn More
Cole-Parmer Cole-Parmer K1VAC Sealed SS Vacuum Transducer, 30-0 "Hg, 4-20 mA Out
Supplier: Cole-Parmer 0735610
Stainless steel diaphragm and polysilicon strain gauge are bonded by vapor deposition—no epoxies or glues which can cause signal instability or drift.