Learn More
Icon Scientific Inc K-7400S Semi-Micro Osmometer
Supplier: Icon Scientific Inc A0006AC
Freezing point osmometer for the determination of osmolality or depressed freezing point of aqueous solutions.