Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific Supplies Jupiter Electronic Pipette
The Jupiter Electronic Pipette has deluxe features at an affordable price.
$563.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Disposable | Reusable |
For Use With | Pipette Tips |
Number of Channels | 1 |
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
S82936
|
United Scientific Supplies
UNEPV0010P |
0.5 to 10 μL |
Each for $563.00
|
|
|||||
S82937
|
United Scientific Supplies
UNEPV0050P |
5 to 50 μL |
Each for $563.00
|
|
|||||
S82938
|
United Scientific Supplies
UNEPV0300P |
30 to 300 μL |
Each for $563.00
|
|
|||||
S82939
|
United Scientific Supplies
UNEPV1000P |
100 to 1000 μL |
Each for $563.00
|
|
|||||
Description
- Versatile United Scientific™ Jupiter Electronic Pipettes provide fast, accurate results using three separate modes: pipetting, stepper dispensing, and diluting.
- Quickly select between modes, volumes, and speed using easy two-knob operation and a digital display with clear status indicators.
- Choose the parameter to be adjusted using the blue knob, and dial in precise settings using the black function wheel.
- Jupiter pipettes use a reliable, high-performance motor to deliver accuracy and repeatability.
- The ergonomic, lightweight design is comfortable to handle, and the electronic operation protects against repetitive motion injury.
- Three separate aspirating and dispensing speeds and up to ten stepper counts let users decide the frequency and quantity of samples to dispense.
- The lithium-ion battery in Jupiter is easily recharged using either the provided AC adapter or USB cable.
- Also includes a stand that can be mounted on a flat surface or wall.
- Electronic operation saves time and reduces manual errors.
- Comfortable, lightweight design.
- Three operating modes with quick, accurate adjustments using two knobs.
- Calibrated and tested in compliance with ISO8655-6 / DIN12650.
Specifications
Not Autoclavable | |
Reusable | |
1 | |
Electronic Pipettor |
White | |
Pipette Tips | |
Non-sterile |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title