Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PREMIUM QUALITY Prepared Microslide: Female Chromosomes
SCI_ED

Smear from blood culture spread in stage of metaphase for individual chromosomes.

Supplier:  Johannes Lieder GmbH & Co. KG HO1041I

Catalog No. S97686

Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.


Includes: Manuals with explanatory comments and designs for each slide

Description

Description

LIEDER high quality microslides are manufactured in rigorous scientific controled laboratory conditions. Slides sets MICR-O-SLIDE are prepared with top quality histologically fixed materials and thus meet all scientific requirements.

  • Best glasses with fine ground edges
  • Size: 26 x 76mm (1 x 3 in.)
  • Furnished with printed labels
  • Packed in solid plastic box

Specifications

Specifications

Type: female chromosome
1 Ea.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit