Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Jade Bottle Top Dispenser
This bottle top dispenser features a recirculation valve.
$460.00 - $734.00
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | ISO 8655 |
Color | White |
Channels | 1 Channel |
Material | PTFE |
Catalog Number | Mfr. No. | Increments | Volume (Metric) | Weight (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Increments | Volume (Metric) | Weight (English) | Price | Quantity & Availability | |||
S139573
|
United Scientific™
BTJD2.5 |
0.05 mL | 0.25 to 2.5 mL | 1.5 lb. |
Each for $460.00
|
|
|||
S139574
|
United Scientific™
BTJD5 |
0.1 mL | 0.5 to 5 mL | 1.5 lb. |
Each for $460.00
|
|
|||
S139575
|
United Scientific™
BTJD10 |
0.2 mL | 1 to 10 mL | 1.75 lb. |
Each for $460.00
|
|
|||
S139576
|
United Scientific™
BTJD30 |
0.5 mL | 2.5 to 30 mL | 2 lb. |
Each for $609.00
|
|
|||
S139577
|
United Scientific™
BTJD60 |
1 mL | 5 to 60 mL | 2.25 lb. |
Each for $609.00
|
|
|||
S139578
|
United Scientific™
BTJD100 |
2 mL | 10 to 100 mL | 4 lb. |
Each for $734.00
|
|
|||
Description
Dispense a wide range of reagents with precision and accuracy using the Jade Bottle Top Dispenser.
- Bottle top dispensers are available in a variety of a volume ranges
- Five adapters are included with each unit for use with a wide range of bottle sizes
- Adapters included are: 28 mm, 32 mm, 38 mm, 40 mm and 45 mm
- Features a recirculation valve that redirects liquid back into the bottle without the loss of reagent
- Operates with a smooth plunger movement for bubble-free dispensing and effortless performance, even under demanding laboratory conditions
- Fully Autoclavable at 121°C, 15 psi for 10 to 15 minutes
Specifications
Autoclavable | |
White | |
PTFE | |
Dispensing Liquids | |
Dispenser and Adapters | |
1 Year |
ISO 8655 | |
1 Channel | |
Analog | |
Chemicals | |
Non-sterile | |
1/Pk. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title