Learn More
McKesson General Medical Suture, Vicryl UD BR CT 4-0 1 8" PC1, 12/pk
Supplier:  McKesson General Medical 124799
Encompass
Suture is indicated for use in general soft tissue approximation and/or ligation, including use in ophthalmic procedures, but not for use in cardiovascular or neurological tissues. Ease of knot tying and knot security on the first throw. Frequent uses general tissue/muscle approximation, ligatures, reproductive tract, ophthalmic, orthopedic (2nd choice). Loss of 50% tensile strength after 3 weeks. Complete mass absorption in 56 to 76 days.
Catalog No. NC1691667
Due to product restrictions, please to purchase or view availability for this product.