Learn More
ITL Corporation SampLok™ Adapter Caps
Used to collect blood culture samples for bacterial detection testing and laboratory tests
$187.00 - $595.00
Specifications
For Use With (Application) | Allow sampling with variety of blood culture bottles and vacuum test tubes |
---|
Catalog Number | Mfr. No. | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sterility | Price | Quantity & Availability | |||||
21-010-223
|
ITL Corporation
A100710 |
Sterile |
Case of 100 for $187.00
|
|
|||||
21-010-222
|
ITL Corporation
A100706 |
Non-sterile |
Case of 350 for $595.00
|
|
|||||
Description
- Available sterile or non-sterile
- Allow sampling with variety of blood culture bottles and vacuum test tubes
- Reduces potential risk of needle stick injury
- With integrated safety lid
- Available with or without pre-attached standard slip luer
- Adapter cap insert available; see cat. no. 21010224
Specifications
Allow sampling with variety of blood culture bottles and vacuum test tubes |