Learn More
Accriva Diagnostics Tenderlett™ Fingerstick Incision Device
Contoured to minimize skin indentation because it matches the shape of the finger
Supplier: Accriva Diagnostics 000TL100I
Description
- Surgical stainless-steel blade creates a small, shallow incision in the finger which cuts more of the capillary bed without cutting too deeply
- Automated manufacturing minimizes variables
- Virtually pain-free: blade permanently retracts after use, providing an added factor of safety for the patient and medical professional
- Nonslip trigger surface prevents the device from slipping once blade-slot surface positioned
- Color coded
Order Info
Available in adult and junior incision depths.
Specifications
100 Pack | |
Fingerstick Incision Device | |
For patient and medical professional |
0.06 in. | |
1.75mm |