Learn More
MP Biomedicals™ Isis DNA Polymerase 100U (1U/ul)
Isis DNA Polymerase 100U (1U/ul)
Supplier:  MP Biomedicals™ 11EPSIS100
 View more versions of this product
Catalog No. MP1EPSIS100
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Ideal for high-fidelity amplification
  • Use to amplify DNA with difficult to read secondary structuresOptimal for blunt-end PCR cloning
  • The most thermostable polymerase available
  • Useful to amplify GC rich sequences
  • Isis™ DNA Polymerase is the most thermostable proofreading polymerase available for high fidelity PCR.
  • Isis DNA Pol Kit contains Isis DNA polymerase, optimized incubation buffers with and without MgCl2 and a separate MgCl2.
Specifications
Specifications
100 u
Product Suggestions
Product Suggestions